missing translation for 'onlineSavingsMsg'
Learn More

N2-Acetylguanine, Mikromol™

Product Code. 30052528
Change view
Click to view available options
Quantity:
25 mg
100 mg
4 x 25 mg
Unit Size:
100mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30052528 100 mg 100mg
30050521 25 mg 25mg
30461836 4 x 25 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30052528 Supplier Mikromol Supplier No. MM0061.12

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Discover Mikromol - your trusted source for high-quality pharmaceutical reference standards. Supporting accurate, compliant analysis with ISO 17034-accredited materials designed for confidence in every result.

N2-Acetylguanine
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N2-Acetylguanine
CAS 19962-37-9
Analyte or Component Names N2-Acetylguanine
Shipping Condition Room Temperature
Molecular Formula C7 H7 N5 O2
Certifications/Compliance ISO 17025
Synonym N-(6,9-Dihydro-6-oxo-1H-purin-2-yl)acetamide, Acetamide, N-(1,6-dihydro-6-oxopurin-2-yl)- (8CI), Acetamide, N-(6,7-dihydro-6-oxo-1H-purin-2-yl)- (9CI), Guanine, N-acetyl- (6CI), 2-Acetylamino-6-hydroxypurine, 2-N-Acetylguanine, N-Acetylguanine, N2-Acetylguanine
InChI Formula InChI=1S/C7H7N5O2/c1-3(13)10-7-11-5-4(6(14)12-7)8-2-9-5/h2H,1H3,(H3,8,9,10,11,12,13,14)
SMILES CC(=O)NC1=Nc2[nH]cnc2C(=O)N1
IUPAC Name N-(6-oxo-1,9-dihydropurin-2-yl)acetamide
Molecular Weight (g/mol) 193.16
Quantity 100 mg
Formula Weight 193.06 g/mol
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.