missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N2,9-Diacetylguanine, TRC
Click to view available options
Quantity:
10 g
Unit Size:
10g
Specifications
Specifications
| Chemical Name or Material | N 2,9-Diacetylguanine |
| CAS | 3056-33-5 |
| Molecular Formula | C9 H9 N5 O3 |
| Quantity | 10 g |
| Synonym | Acyclovir Impurity L |
| InChI Formula | InChI=1S/C9H9N5O3/c1-4(15)11-9-12-7-6(8(17)13-9)10-3-14(7)5(2)16/h3H,1-2H3,(H2,11,12,13,15,17) |
| SMILES | CC(=O)NC1=Nc2c(ncn2C(=O)C)C(=O)N1 |
| IUPAC Name | N-(9-acetyl-6-oxo-1H-purin-2-yl)acetamide |
| Molecular Weight (g/mol) | 235.2 |
| Formula Weight | 235.0705 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction