missing translation for 'onlineSavingsMsg'
Learn More

N2-[(3R)-3-Hydroxy-1-oxododecyl]-L-asparaginyl-N1-[(1S)-1-(diethoxymethyl)-3-methylbutyl]-L-glutamamide, TRC

Product Code. 30393340
Change view
Click to view available options
Quantity:
1 mg
2.5 mg
5 mg
Unit Size:
2.5mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30393340 5 mg -
30401091 1 mg -
30406145 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 30393340 Supplier Toronto Research Chemicals Supplier No. TRCH1370005MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1777816-46-2
Molecular Formula C31H59N5O8
Molecular Weight (g/mol) 629.83
IUPAC Name (2S)-2-[[(2S)-4-amino-2-[[(3R)-3-hydroxydodecanoyl]amino]-4-oxobutanoyl]amino]-N-(1,1-diethoxy-4-methylpentan-2-yl)pentanediamide
SMILES CCCCCCCCC[C@@H](O)CC(=O)N[C@@H](CC(=O)N)C(=O)N[C@@H](CCC(=O)N)C(=O)NC(CC(C)C)C(OCC)OCC

Specifications

Quantity 5 mg
InChI Formula InChI=1S/C31H59N5O8/c1-6-9-10-11-12-13-14-15-22(37)19-28(40)34-24(20-27(33)39)30(42)35-23(16-17-26(32)38)29(41)36-25(18-21(4)5)31(43-7-2)44-8-3/h21-25,31,37H,6-20H2,1-5H3,(H2,32,38)(H2,33,39)(H,34,40)(H,35,42)(H,36,41)/t22-,23+,24+,25?/m1/s1
Formula Weight 629.4364
Chemical Name or Material N2-[(3R)-3-Hydroxy-1-oxododecyl]-L-asparaginyl-N1-[(1S)-1-(diethoxymethyl)-3-methylbutyl]-L-glutamamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.