Learn More
Macherey-Nagel™ N13 Aluminum Closure
Product Code. 10047640 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Gold
Unit Size:
Pack of 100
Product Code. Color unitSize
10047640 Gold Pack of 100
1 options
Product Code. 10047640 Supplier Macherey-Nagel™ Supplier No. 70232
Only null left
Add to basket
Add to basket
Specifications
Color Gold
Material Aluminum, Butyl Rubber, PTFE
Septum Butyl dark grey / PTFE grey(only centrically laminated, typically called Pharma-Fix)
Quantity 100 Pk.
Thickness (Metric) 2 mm
Product Type Center Tear-off