Learn More
Macherey-Nagel™ N11 Aluminum Closure
Product Code. 10043197 Shop All Macherey-Nagel Products
Change view
Click to view available options
Color:
Silver
Unit Size:
Pack of 100
Product Code. Color unitSize
10043197 Silver Pack of 100
1 options
Product Code. 10043197 Supplier Macherey-Nagel™ Supplier No. 702730
Only null left
Add to basket
Add to basket
Specifications
Color Silver
Material Aluminum, FEP, Rubber
Septum Red Rubber / FEP colorless
Quantity 100 Pk.
Thickness (Metric) 1 mm
Product Type Crimp Cap