missing translation for 'onlineSavingsMsg'
Learn More

N1,N4-Diphenylbenzene-1,4-diamine, TRC

Product Code. 30385320
Change view
Click to view available options
Quantity:
500 mg
Unit Size:
500mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30385320 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30385320 Supplier Toronto Research Chemicals Supplier No. TRCD493428500MG
 Restricted Item
Estimated Shipment: 12-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N,N'-Diphenyl-p-phenylenediamine
CAS 74-31-7
Molecular Formula C18 H16 N2
Quantity 500 mg
Synonym 1,4-Benzenediamine, N1,N4-diphenyl-, 1,4-Benzenediamine, N,N'-diphenyl- (9CI), p-Phenylenediamine, N,N'-diphenyl- (8CI), N1,N4-Diphenyl-1,4-benzenediamine, 1,4-Bis(phenylamino)benzene, 1,4-Dianilinobenzene, 4,4'-Diphenyl-p-phenylenediamine, 4-Phenylaminodiphenylamine, AgeRite DPPD, Altofane DIP, Antage DP, Antigen P, Antigene P, Antioxidant H, DFFD, DPPD, Diafen FF, Diafen FF-T, Diphenyl-p-phenylenediamine, Ekaland DPPD, Flexamine G, JZF, N,N'-Diphenyl-1,4-benzenediamine, N,N'-Diphenyl-1,4-phenylenediamine, N,N'-Diphenyl-p-phenylenediamine, NSC 5761, Naugard J, Nocrac DP, Nonflex H, Nonox DPPD, PPD, Permanax 18, Permanax DPPD, Stabilizer DPPD, p-Bis(phenylamino)benzene, p-Phenylaminodiphenylamine
InChI Formula InChI=1S/C18H16N2/c1-3-7-15(8-4-1)19-17-11-13-18(14-12-17)20-16-9-5-2-6-10-16/h1-14,19-20H
SMILES N(c1ccccc1)c2ccc(Nc3ccccc3)cc2
IUPAC Name 1-N,4-N-diphenylbenzene-1,4-diamine
Molecular Weight (g/mol) 260.33
Formula Weight 260.1313
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.