missing translation for 'onlineSavingsMsg'
Learn More

N1-(5-Oxo-5,6,7,8-tetrahydronaphthalen-2-yl)acetamide, 97%, Thermo Scientific™

Product Code. 10590922
Change view
Click to view available options
Quantity:
10 g
Packaging:
Amber Glass Bottle
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10590922 10 g Amber Glass Bottle 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10590922 Supplier Thermo Scientific Maybridge Supplier No. NRB00998EA
 Restricted Item
Estimated Shipment: 04-11-2026
Add to basket
This item is not returnable. View return policy

Specifications

CAS 88611-67-0
CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Molecular Formula C12H13NO2
MDL Number MFCD00099461
Quantity 10 g
Synonym n-5-oxo-5,6,7,8-tetrahydronaphthalen-2-yl acetamide, n1-5-oxo-5,6,7,8-tetrahydronaphthalen-2-yl acetamide, 6-acetamido-1,2,3,4-tetrahydronaphthalen-1-one, n-5-oxo-7,8-dihydro-6h-naphthalen-2-yl acetamide, acetamide,n-5,6,7,8-tetrahydro-5-oxo-2-naphthalenyl, acmc-20lbzu, 6-acetamido-tetralone, pubchem17583, 6-acetamido-1-tetralone, maybridge1_005781
InChI Key WEBCZGJWXXPNHB-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC2=C(C=C1)C(=O)CCC2
IUPAC Name N-(5-oxo-7,8-dihydro-6H-naphthalen-2-yl)acetamide
Molecular Weight (g/mol) 203.241
PubChem CID 2780190
Formula Weight 203.24
Percent Purity 97%
Chemical Name or Material N1-(5-oxo-5,6,7,8-tetrahydronaphthalen-2-yl)acetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.