missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N1-(4-pyridyl)benzamide, Thermo Scientific™
Click to view available options
Quantity:
10 g
25 g
50 g
Specifications
CAS Min % | 97.0 |
Assay Percent Range | 97% |
Packaging | Amber glass bottle |
Quantity | 50 g |
Formula Weight | 198.22 |
Percent Purity | 97% |
Chemical Name or Material | N1-(4-Pyridyl)benzamide |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'