missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-pyridyl)benzamide, Thermo Scientific™

Product Code. 10396714
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10396714 25 g 25g
10065492 10 g 10g
11326246 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10396714 Supplier Thermo Scientific Maybridge Supplier No. NRB04027EB
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5221-44-3
Molecular Formula C12H10N2O
Molecular Weight (g/mol) 198.225
MDL Number MFCD00160355
InChI Key PNWVOLKZHJEWQU-UHFFFAOYSA-N
Synonym n-pyridin-4-yl benzamide, 4-benzamidopyridine, n1-4-pyridyl benzamide, n-4-pyridylbenzamide, 4-benzoylaminopyridine, pyridine, 4-benzamido, n-4-pyridyl benzamide, benzamide, n-4-pyridinyl-9ci, n-benzoyl-4-aminopyridine, phenyl-n-4-pyridyl carboxamide
PubChem CID 78893
IUPAC Name N-pyridin-4-ylbenzamide
SMILES C1=CC=C(C=C1)C(=O)NC2=CC=NC=C2

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Quantity 25 g
Formula Weight 198.22
Percent Purity 97%
Chemical Name or Material N1-(4-Pyridyl)benzamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.