missing translation for 'onlineSavingsMsg'
Learn More

N1-[4-Cyano-2-(trifluoromethoxy)phenyl]acetamide, ≥95%, Thermo Scientific™

Product Code. p-2705648
Click to view available options
Quantity:
1 g
10 g
25 g
Unit Size:
10g
1g
25g
This item is not returnable. View return policy

Product Code. 10180892

Brand: Thermo Scientific Maybridge TL00674DA

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 175278-19-0
Molecular Formula C10H7F3N2O2
Molecular Weight (g/mol) 244.173
MDL Number MFCD00204174
InChI Key RHBNAXXJVYFEEA-UHFFFAOYSA-N
Synonym n-4-cyano-2-trifluoromethoxy phenyl acetamide, 4-cyano-2-trifluoromethoxy acetanilide, n1-4-cyano-2-trifluoromethoxy phenyl acetamide, 4'-cyano-2'-trifluoromethoxy acetanilide, maybridge1_000083, 4'-cyano-2'-trifluoromethoxy acetanilid, n-4-cyano-2-trifluoromethoxyphenyl-acetamide
PubChem CID 2736739
IUPAC Name N-[4-cyano-2-(trifluoromethoxy)phenyl]acetamide
SMILES CC(=O)NC1=C(C=C(C=C1)C#N)OC(F)(F)F

Specifications

CAS Min % 95.0
Assay Percent Range 95%+
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 244.17
Percent Purity 95%
Chemical Name or Material N1-[4-Cyano-2-(trifluoromethoxy)phenyl]acetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.