missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Butyryl-3-hydroxyphenyl)acetamide, Tech., Thermo Scientific™

Product Code. 10006374
Change view
Click to view available options
Quantity:
1 g
5 g
10 g
Unit Size:
10g
1g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10006374 5 g 5g
10495132 1 g 1g
10049571 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10006374 Supplier Thermo Scientific Maybridge Supplier No. SB01739DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 28583-62-2
Molecular Formula C12H15NO3
Molecular Weight (g/mol) 221.26
MDL Number MFCD00276573
InChI Key RSBIKRQPSYDRFZ-UHFFFAOYSA-N
Synonym n1-4-butyryl-3-hydroxyphenyl acetamide, unii-4kn634aqj7, n-4-butanoyl-3-hydroxyphenyl acetamide, 4'-butyryl-3'-hydroxyacetanilide, n-4-butyryl-3-hydroxyphenyl acetamide, n-3-hydroxy-4-1-oxobutyl phenyl acetamide, maybridge3_006119, acetamide,n-3-hydroxy-4-1-oxobutyl phenyl, acetamide, n-3-hydroxy-4-1-oxobutyl phenyl
PubChem CID 119989
IUPAC Name N-(4-butanoyl-3-hydroxyphenyl)acetamide
SMILES CCCC(=O)C1=CC=C(NC(C)=O)C=C1O

Specifications

Quantity 5 g
Formula Weight 221.25
Percent Purity Tech.
Grade Technical
Chemical Name or Material N1-(4-Butyryl-3-hydroxyphenyl)acetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.