missing translation for 'onlineSavingsMsg'
Learn More

N1-[4-Bromo-2-(trifluoromethoxy)phenyl]acetamide, 97%, Thermo Scientific™

Product Code. 10120852
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
10120852 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10120852 Supplier Thermo Scientific Maybridge Supplier No. TL00670DA
 Restricted Item
Estimated Shipment: 25-01-2027
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 175278-18-9
CAS Min % 97.0
Assay Percent Range 97%
Molecular Formula C9H7BrF3NO2
MDL Number MFCD00203979
Quantity 1 g
Synonym n-4-bromo-2-trifluoromethoxy phenyl acetamide, 4-bromo-2-trifluoromethoxy acetanilide, 4'-bromo-2'-trifluoromethoxy acetanilide, 1-acetamido-4-bromo-2-trifluoromethoxy benzene, n1-4-bromo-2-trifluoromethoxy phenyl acetamide, maybridge1_000061, acmc-209e9r, 4-bromo-2-trifluoromethyl acetanilide
InChI Key SHHWWZMWTUTJPY-UHFFFAOYSA-N
SMILES CC(=O)NC1=CC=C(Br)C=C1OC(F)(F)F
IUPAC Name N-[4-bromo-2-(trifluoromethoxy)phenyl]acetamide
Molecular Weight (g/mol) 298.06
PubChem CID 2736416
Formula Weight 298.06
Percent Purity 97%
Chemical Name or Material N1-[4-Bromo-2-(trifluoromethoxy)phenyl]acetamide
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.