missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Thermo Scientific™

Product Code. p-4511391
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10442633

Brand: Thermo Scientific Maybridge BTB13402CB

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 119-63-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
InChI Key QFELUFGHFLYZEZ-UHFFFAOYSA-N
Synonym n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
PubChem CID 67068
IUPAC Name N-(4-aminophenyl)-N-methylacetamide
SMILES CC(=O)N(C)C1=CC=C(C=C1)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 164.21
Percent Purity Tech..
Grade Technical
Chemical Name or Material N1-(4-Aminophenyl)-N1-methylacetamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.