missing translation for 'onlineSavingsMsg'
Learn More

N1-(4-Aminophenyl)-N1-methylacetamide, 97%, Thermo Scientific™

Product Code. 10651226
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10651226 5 g 5g
10442633 250 mg 250mg
10749793 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10651226 Supplier Thermo Scientific Maybridge Supplier No. BTB13402DE
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 119-63-1
Molecular Formula C9H12N2O
Molecular Weight (g/mol) 164.208
InChI Key QFELUFGHFLYZEZ-UHFFFAOYSA-N
Synonym n-4-aminophenyl-n-methylacetamide, 4'-amino-n-methylacetanilide, p-amino-n-methylacetanilide, acetamide, n-4-aminophenyl-n-methyl, n1-4-aminophenyl-n1-methylacetamide, 4-amino-n-acetyl-n-methylaniline, n-4-aminophenyl-n-methyl-acetamide, p-amino-n-acetyl-n-methylaniline, acetanilide, 4'-amino-n-methyl, 4-amino-n-methyl acetanilide
PubChem CID 67068
IUPAC Name N-(4-aminophenyl)-N-methylacetamide
SMILES CC(=O)N(C)C1=CC=C(C=C1)N

Specifications

Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 164.21
Percent Purity Tech..
Grade Technical
Chemical Name or Material N1-(4-Aminophenyl)-N1-methylacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.