missing translation for 'onlineSavingsMsg'
Learn More

N1-(2-Chlorophenyl)-2-bromoacetamide, 95%, Thermo Scientific™

Product Code. 10101042
Change view
Click to view available options
Quantity:
1 g
10 g
Unit Size:
10g
1g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10101042 10 g 10g
10191032 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10101042 Supplier Thermo Scientific Maybridge Supplier No. SPB05477EA
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 5439-11-2
Molecular Formula C8H7BrClNO
Molecular Weight (g/mol) 248.50
MDL Number MFCD00173815
InChI Key WIOJXHICOOQVAB-UHFFFAOYSA-N
Synonym 2-bromo-n-2-chlorophenyl acetamide, n1-2-chlorophenyl-2-bromoacetamide, n-bromoacetyl-2'-chloroacetaniline, wiojxhicooqvab-uhfffaoysa-n, 2-bromo-n-2-chloro-phenyl-acetamide, acetamide, 2-bromo-n-2-chlorophenyl
PubChem CID 225525

Specifications

CAS Min % 97.0
Assay Percent Range 95%
Linear Formula ClC6H4NHC(O)CH2Br
Quantity 10 g
Formula Weight 248.51
Percent Purity 95%
Chemical Name or Material N1-(2-chlorophenyl)-2-bromoacetamide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.