missing translation for 'onlineSavingsMsg'
Learn More

[N(Z)]-N-(3,5-Dimethyl-2(3H)-thiazolylidene)-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, TRC

Product Code. 30380344
Change view
Click to view available options
Quantity:
10 mg
100 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30380344 10 mg
30379294 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30380344 Supplier Toronto Research Chemicals Supplier No. TRCD47947510MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1262333-25-4
Molecular Formula C15 H15 N3 O4 S2
Molecular Weight (g/mol) 365.427
Synonym N-[(2Z)-3,5-Dimethylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide, Meloxicam Imp. C (EP), (Z)-N'-Methylmeloxicam, (Z)-N-(3,5-dimethylthiazol-2(3H)-ylidene)-4-hydroxy-2-methyl-2H-benzo[e][1,2]thiazine-3-carboxamide 1,1-dioxide
IUPAC Name (NZ)-N-(3,5-dimethyl-1,3-thiazol-2-ylidene)-4-hydroxy-2-methyl-1,1-dioxo-1λ^{6},2-benzothiazine-3-carboxamide
SMILES CN1C=C(C)S/C/1=N\C(=O)C2=C(O)c3ccccc3S(=O)(=O)N2C

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C15H15N3O4S2/c1-9-8-17(2)15(23-9)16-14(20)12-13(19)10-6-4-5-7-11(10)24(21,22)18(12)3/h4-8,19H,1-3H3/b16-15-
Formula Weight 365.05
Chemical Name or Material N-[(2Z)-3,5-Dimethylthiazol-2(3H)-ylidene]-4-hydroxy-2-methyl-2H-1,2-benzothiazine-3-carboxamide 1,1-Dioxide
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.