missing translation for 'onlineSavingsMsg'
Learn More

N^w-Nitro-L-arginine, 98%

Product Code. 15448648
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15448648 25 g 25g
15438648 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15448648 Supplier Thermo Scientific Alfa Aesar Supplier No. H62061.14
 Restricted Item
Estimated Shipment: 09-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 2149-70-4 | C6H13N5O4 | 219.201 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 2149-70-4
Molecular Formula C6H13N5O4
Molecular Weight (g/mol) 219.201
MDL Number MFCD00007033
InChI Key MRAUNPAHJZDYCK-BYPYZUCNSA-N
Synonym h-arg no2-oh, ng-nitro-l-arginine, l-nna, nitroarginine, n-nitro-l-arginine, nomega-nitro-l-arginine, n g-nitro-l-arginine, ng-no2-l-arg, n-omega-nitro-l-arginine, n g-nitroarginine
PubChem CID 440005
ChEBI CHEBI:27960
SMILES C(CC(C(=O)O)N)CN=C(N)N[N+](=O)[O-]

Specifications

Melting Point 257°C
Beilstein 1728914
Quantity 25 g
Formula Weight 219.2
Percent Purity 98%
Chemical Name or Material Nomega-Nitro-L-arginine

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.