missing translation for 'onlineSavingsMsg'
Learn More

N-Vinyl-2-pyrrolidone, 99%, stabilized with Kerobit(R), Thermo Scientific Chemicals

Product Code. 11946621 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
2.5 L
10 L
Packaging:
Glass Bottle
Plastic drum
Unit Size:
10 L
2.5 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
11946621 10 L Plastic drum 10 L
10648444 2.5 L Glass Bottle 2.5 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11946621 Supplier Thermo Scientific Acros Supplier No. 354730100
 Restricted Item
Estimated Shipment: 07-12-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 88-12-0 | C6H9NO | 111.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88-12-0
Molecular Formula C6H9NO
Molecular Weight (g/mol) 111.14
InChI Key WHNWPMSKXPGLAX-UHFFFAOYSA-N
Synonym n-vinyl-2-pyrrolidone, n-vinylpyrrolidone, 1-vinyl-2-pyrrolidone, 1-vinylpyrrolidin-2-one, n-vinyl-2-pyrrolidinone, vinylpyrrolidone, 2-pyrrolidinone, 1-ethenyl, 1-vinyl-2-pyrrolidinone, n-vinylpyrrolidinone, 1-vinylpyrrolidone
PubChem CID 6917
ChEBI CHEBI:82551
IUPAC Name 1-ethenylpyrrolidin-2-one
SMILES C=CN1CCCC1=O

Specifications

Melting Point 13.0°C to 14.0°C
Color Colorless to Yellow
Density 1.0430g/mL
Boiling Point 218.0°C
Flash Point 95°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Plastic drum
Refractive Index 1.5119 to 1.5139
Quantity 10 L
Merck Index 15,7814
Specific Gravity 1.043
Solubility Information Solubility in water: 52.1g/L (25°C). Other solubilities: miscible with acetone,diethyl ether,ethanol,,toluene,benzene,very good soluble in organic solvents
Stabilizer 10ppm kerobit
Viscosity 2.4 mPa.s (20°C)
Formula Weight 111.14
Percent Purity 99%
Physical Form Liquid
Chemical Name or Material N-Vinyl-2-pyrrolidone
Show More Show Less
compliance-icons compliance-icons compliance-icons
Product Identifier
  • N-Vinyl-2-pyrrolidone, stabilized
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity after repeated exposure Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H318-Causes serious eye damage.
  • H351-Suspected of causing cancer.
  • H373-May cause damage to organs.
  • H335-May cause respiratory irritation.
  • H312-Harmful in contact with skin.
  • H332-Harmful if inhaled.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.