missing translation for 'onlineSavingsMsg'
Learn More

n-Undecanoyl chloride, 98%

Product Code. 15442848
Change view
Click to view available options
Quantity:
500 mg
1 g
Unit Size:
1g
500mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15442848 500 mg 500mg
15432848 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15442848 Supplier Thermo Scientific Alfa Aesar Supplier No. H56380.ME
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 17746-05-3 | C11H21ClO | 204.738 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 17746-05-3
Molecular Formula C11H21ClO
Molecular Weight (g/mol) 204.738
MDL Number MFCD00000739
InChI Key JUKPJGZUFHCZQI-UHFFFAOYSA-N
Synonym n-undecanoyl chloride, undecanoic acid chloride, undecanoylchloride, n-undecanoylchloride, undecanoyl chloride, acmc-1c80a, ksc175q6f
PubChem CID 87287
IUPAC Name undecanoyl chloride
SMILES CCCCCCCCCCC(=O)Cl

Specifications

Boiling Point 135°C to 136°C
Flash Point 113°C (235°F)
Linear Formula CH3(CH2)9COCl
Refractive Index 1.443
Quantity 500 mg
UN Number UN3265
Beilstein 1759226
Sensitivity Moisture sensitive
Formula Weight 204.74
Percent Purity 98%
Chemical Name or Material n-Undecanoyl chloride
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.