missing translation for 'onlineSavingsMsg'
Learn More

n-Undecane, 99%

Product Code. p-210818 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
100 mL
500 mL
2.5 L
10 L
Unit Size:
10 L
100mL
2.5 L
500mL
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 10501201

Brand: Thermo Scientific Acros 140661000

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 1120-21-4 | C11H24 | 156.31 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1120-21-4
Molecular Formula C11H24
Molecular Weight (g/mol) 156.31
MDL Number MFCD00008959
InChI Key RSJKGSCJYJTIGS-UHFFFAOYSA-N
Synonym n-undecane, hendecane, n-hendecane, decane, methyl, unii-jv0qt00nue, ccris 3796, jv0qt00nue, undecane, undecane, analytical standard, und
PubChem CID 14257
ChEBI CHEBI:46342
IUPAC Name undecane
SMILES CCCCCCCCCCC

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.7400g/mL
Infrared Spectrum Authentic
Assay Percent Range 99%
Linear Formula CH3(CH2)9CH3
Refractive Index 1.4160 to 1.4180 (20°C, 589nm)
Quantity 100 mL
Beilstein 01, IV, 487
Formula Weight 156.31
Vapor Pressure 0.6hPa at 20°C
Percent Purity ≥98.5%
Physical Form Liquid
Chemical Name or Material n-Undecane, 99%
Show More Show Less
compliance-icons
Product Identifier
  • n-Undecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P331-Do NOT induce vomiting.

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.