missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-Trityl Candesartan Methyl Ester-d4, TRC
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
Specifications
Specifications
| Chemical Name or Material | N-Trityl Candesartan Methyl Ester-d4 |
| Molecular Formula | C44 D4 H32 N6 O3 |
| Quantity | 25 mg |
| Synonym | 2-Ethoxy-1-[[2'-[2-(triphenylmethyl)-2H-tetrazol-5-yl][1,1'-biphenyl]-4-yl]methyl]-1H-benzimidazole-7-carboxylic Acid Methyl Ester-d4, 2-Ethoxy-1-[(2'-(2-trityl-2H-tetrazol-5-yl)-1,1'-biphenyl-4-yl)methyl]-7-benzimidazolecarboxylic Acid Methyl Ester-d4 |
| InChI Formula | InChI=1S/C44H36N6O3/c1-3-53-43-45-39-25-15-24-38(42(51)52-2)40(39)49(43)30-31-26-28-32(29-27-31)36-22-13-14-23-37(36)41-46-48-50(47-41)44(33-16-7-4-8-17-33,34-18-9-5-10-19-34)35-20-11-6-12-21-35/h4-29H,3,30H2,1-2H3/i26D,27D,28D,29D |
| SMILES | [2H]c1c([2H])c(c([2H])c([2H])c1Cn2c(OCC)nc3cccc(C(=O)OC)c23)c4ccccc4c5nnn(n5)C(c6ccccc6)(c7ccccc7)c8ccccc8 |
| IUPAC Name | methyl 2-ethoxy-3-[[2,3,5,6-tetradeuterio-4-[2-(2-trityltetrazol-5-yl)phenyl]phenyl]methyl]benzimidazole-4-carboxylate |
| Molecular Weight (g/mol) | 700.82 |
| Formula Weight | 700.31 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction