missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Specifications
Specifications
| CAS | 4-1-5704 |
| Melting Point | 182°C |
| Color | White |
| pH | 4.8 to 5.4 |
| Packaging | Plastic powder jar |
| Molecular Formula | C6H13NO5 |
| MDL Number | 4277 |
| Quantity | 100 g |
| Synonym | tricine, n-tris hydroxymethyl methylglycine, n-tri hydroxymethyl methyl glycine, n-tris hydroxymethyl methyl glycine, glycine, n-2-hydroxy-1,1-bis hydroxymethyl ethyl, n-2-hydroxy-1,1-bis hydroxymethyl ethyl glycine, unii-w12lh4v8v3, n-tris-hydroxymethyl methylglycine |
| InChI Key | SEQKRHFRPICQDD-UHFFFAOYSA-N |
| Show More |
Safety and Handling
Product Identifier
- Tricine
- Warning
- Serious eye damage/eye irritation Category 2
- Skin corrosion/irritation Category 2
- Specific target organ toxicity Category 3
- H319-Causes serious eye irritation.
- H335-May cause respiratory irritation.
- H315-Causes skin irritation.
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?