missing translation for 'onlineSavingsMsg'
Learn More

N-Tris(Hydroxymethyl)Methyl-2-Aminoethanesulfonic Acid, for HPLC, Biological Buffer, Fisher Chemical

Product Code. 10795341 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
Packaging:
Plastic powder jar
Unit Size:
100g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10795341 100 g Plastic powder jar 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10795341 Supplier Fisher Chemical Supplier No. T/3720/48

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Specifications

CAS 7365-44-8
Melting Point 223°C
Color White
pH 4.0 to 4.8
Packaging Plastic powder jar
Molecular Formula C6H15NO6S
MDL Number 7532
Quantity 100 g
Synonym tes, n-tris hydroxymethyl methyl-2-aminoethanesulfonic acid, tes buffer, tes free acid, ethanesulfonic acid, 2-2-hydroxy-1,1-bis hydroxymethyl ethyl amino, 2-1,3-dihydroxy-2-hydroxymethyl propan-2-yl amino ethanesulfonic acid, 2-2-hydroxy-1,1-dihydroxymethyl-ethylamino-ethanesulfonic acid, tris hydroxymethyl methyl-2-aminomethane sulfonic acid, 2-tris hydroxymethyl methylamino ethane-1-sulphonic acid, n-tri hydroxymethyl methyltaurine
InChI Key JOCBASBOOFNAJA-UHFFFAOYSA-N
SMILES C(CS(=O)(=O)O)NC(CO)(CO)CO
IUPAC Name 2-[[1,3-dihydroxy-2-(hydroxymethyl)propan-2-yl]amino]ethanesulfonic acid
Molecular Weight (g/mol) 229.247
PubChem CID 81831
ChEBI CHEBI:44356
Formula Weight 229.25g/mol
Physical Form Powder
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.