missing translation for 'onlineSavingsMsg'
Learn More

N-trifluoroacetyl Varenicline, TRC

Product Code. 30455972
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. Quantity unitSize
30455972 100 mg 100mg
30456751 50 mg -
30456941 500 mg 500mg
3 options
This item is not returnable. View return policy

Product Code. 30455972

Brand: Toronto Research Chemicals TRCT889503100MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 230615-70-0
Formula Weight 307.0932
InChI Formula InChI=1S/C15H12F3N3O/c16-15(17,18)14(22)21-6-8-3-9(7-21)11-5-13-12(4-10(8)11)19-1-2-20-13/h1-2,4-5,8-9H,3,6-7H2
IUPAC Name 2,2,2-trifluoro-1-(5,8,14-triazatetracyclo[10.3.1.02,11.04,9]hexadeca-2,4,6,8,10-pentaen-14-yl)ethanone
Molecular Formula C15 H12 F3 N3 O
Molecular Weight (g/mol) 307.2705
SMILES FC(F)(F)C(=O)N1CC2CC(C1)c3cc4nccnc4cc23
Synonym Ethanone, 2,2,2-trifluoro-1-(6,7,9,10-tetrahydro-6,10-methano-8H-pyrazino[2,3-h][3]benzazepin-8-yl)-, 6,10-Methano-6H-pyrazino[2,3-h][3]benzazepine, 7,8,9,10-tetrahydro-8-(trifluoroacetyl)- (9CI), 2,2,2-Trifluoro-1-(6,7,9,10-tetrahydro-6,10-methano-8H-pyrazino[2,3-h][3]benzazepin-8-yl)ethanone
Chemical Name or Material Varenicline Impurity 16

Specifications

Quantity 100 mg
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.