missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecanoic acid, 97%

Product Code. 10715811 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
10 g
25 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
25g
50g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10715811 25 g Glass bottle 25g
10235190 10 g Glass bottle 10g
10550584 50 g Glass bottle 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10715811 Supplier Thermo Scientific Acros Supplier No. 139520250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-53-9 | C13H26O2 | 214.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-53-9
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00002741
InChI Key SZHOJFHSIKHZHA-UHFFFAOYSA-N
Synonym n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16
PubChem CID 12530
ChEBI CHEBI:45919
IUPAC Name tridecanoic acid
SMILES CCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color White to Yellow
Boiling Point 236°C (100.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3(CH2)11CO2H
Quantity 25 g
Beilstein 02, 364
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 214.35
Percent Purity 97%
Physical Form Crystalline Chunks, Powder or Flakes
Chemical Name or Material n-Tridecanoic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.