missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecanoic acid, 97%

Product Code. 10235190 Shop All Thermo Scientific Chemicals Products
Click to view available options
Quantity:
10 g
25 g
50 g
Packaging:
Glass bottle
Unit Size:
10g
25g
50g
This item is not returnable. View return policy

Product Code. 10235190

Brand: Thermo Scientific Acros 139520100

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 638-53-9 | C13H26O2 | 214.35 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-53-9
Molecular Formula C13H26O2
Molecular Weight (g/mol) 214.35
MDL Number MFCD00002741
InChI Key SZHOJFHSIKHZHA-UHFFFAOYSA-N
Synonym n-tridecanoic acid, tridecylic acid, n-tridecoic acid, unii-19936liy2v, anion, tridecanoic acid, wln: qv12, c10-16 fatty acids, fatty acids, c10-16, carboxylic acids, c10-16
PubChem CID 12530
ChEBI CHEBI:45919
IUPAC Name tridecanoic acid
SMILES CCCCCCCCCCCCC(=O)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 40°C to 45°C
Color White to Yellow
Boiling Point 236°C (100.0 mmHg)
Flash Point 113°C
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass bottle
Linear Formula CH3(CH2)11CO2H
Quantity 10 g
Beilstein 02, 364
Solubility Information Solubility in water: slightly soluble. Other solubilities: soluble in alcohol and ether
Formula Weight 214.35
Percent Purity 97%
Physical Form Crystalline Chunks, Powder or Flakes
Chemical Name or Material n-Tridecanoic acid, 97%
Show More Show Less

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.