missing translation for 'onlineSavingsMsg'
Learn More

n-Tridecane, 98+%

Product Code. 11483747
Click to view available options
Quantity:
25 g
100 g
500 g
Unit Size:
100g
25g
500g
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

Product Code. 11483747

Brand: Thermo Scientific Alfa Aesar A18220.22

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 629-50-5 | C13H28 | 184.37 g/mol

n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tridecane is used as a distillation chaser. It is a component of various fuels and solvents used in industry.

Solubility
Immiscible with water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-50-5
Molecular Formula C13H28
Molecular Weight (g/mol) 184.37
MDL Number MFCD00008979
InChI Key IIYFAKIEWZDVMP-UHFFFAOYSA-N
Synonym n-tridecane, tridekan, alkanes, c12-14, dodecane, methyl, unii-a3lzf0l939, tridecane, n, trd, tridecane, analytical standard, tridecan, tridecyl group
PubChem CID 12388
ChEBI CHEBI:35998
IUPAC Name tridecane
SMILES CCCCCCCCCCCCC

Specifications

Melting Point -5°C
Density 0.755
Boiling Point 235°C
Flash Point 79°C (174°F)
Linear Formula CH3(CH2)11CH3
Refractive Index 1.425
Quantity 100 g
Beilstein 1733089
Solubility Information Immiscible with water.
Formula Weight 184.37
Percent Purity ≥98%
Chemical Name or Material n-Tridecane
Show More Show Less
compliance-icons
Product Identifier
  • n-Tridecane
Signal Word
  • Danger
Hazard Category
  • Aspiration toxicity Category 1
Hazard Statement
  • H304-May be fatal if swallowed and enters airways.
  • EUH066-Repeated exposure may cause skin dryness or cracking.
Precautionary Statement
  • P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
  • P331-Do NOT induce vomiting.
Supplemental information
  • MIXTURE LIST-Contains : Tridecane

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.