missing translation for 'onlineSavingsMsg'
Learn More

n-Tricosane, 99%

Product Code. 11431978
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11431978 5 g 5g
11441978 25 g 25g
11451978 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11431978 Supplier Thermo Scientific Alfa Aesar Supplier No. A14577.06
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 638-67-5 | C23H48 | 324.637 g/mol

n-Tricosane is used as a pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Tricosane is used as a pharmaceutical intermediate.

Solubility
Insoluble in water.

Notes
Store in cool, dry conditions in a well sealed container. Incompatible with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-67-5
Molecular Formula C23H48
Molecular Weight (g/mol) 324.637
MDL Number MFCD00009350
InChI Key FIGVVZUWCLSUEI-UHFFFAOYSA-N
Synonym n-tricosane, tricosan, docosane, methyl, unii-t166b8r1vc, ch3-ch2 21-ch3, tricosane, analytical standard, tricosane, tricosane standardmaterialforgc, unii-j3n6x3yk96 component
PubChem CID 12534
ChEBI CHEBI:32934
IUPAC Name tricosane
SMILES CCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 47°C to 50°C
Density 0.7969
Boiling Point 199°C to 200°C (3 mmHg)
Flash Point >110°C (230°F)
Refractive Index 1.4468
Quantity 5 g
Beilstein 1756000
Solubility Information Insoluble in water.
Formula Weight 324.64
Percent Purity 99%
Chemical Name or Material n-Tricosane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.