missing translation for 'onlineSavingsMsg'
Learn More

n-Triacontane, 98+%

Product Code. 11377425
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11377425 250 mg 250mg
11357425 1 g 1g
11367425 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11377425 Supplier Thermo Scientific Alfa Aesar Supplier No. L05386.MD
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 638-68-6 | C30H62 | 422.826 g/mol

n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
n-Triacontane is used to improve the threshold voltage and mobility for electrons of ambipolar organic light emitting field effect transistors with modified asymmetric electrodes.

Solubility
Soluble in benzene, chloroform and toluene. Insoluble in water.

Notes
Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 638-68-6
Molecular Formula C30H62
Molecular Weight (g/mol) 422.826
MDL Number MFCD00009410
InChI Key JXTPJDDICSTXJX-UHFFFAOYSA-N
Synonym n-triacontane, triacontane, analytical standard, triacontane, acmc-1b4uf, n-triacontane 250mg, ch3-ch2 28-ch3
PubChem CID 12535
ChEBI CHEBI:31006
IUPAC Name triacontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 65°C to 68°C
Boiling Point 455°C
Flash Point 237°C (458°F)
Odor Odorless
Quantity 250 mg
Beilstein 1777281
Solubility Information Soluble in benzene,chloroform and toluene. Insoluble in water.
Formula Weight 422.83
Percent Purity 99%
Chemical Name or Material n-Triacontane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.