missing translation for 'onlineSavingsMsg'
Learn More

N-Thionylaniline, 96%

Product Code. 15465778
Change view
Click to view available options
Quantity:
25 g
Unit Size:
25g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15465778 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15465778 Supplier Thermo Scientific Alfa Aesar Supplier No. H59451.14
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 1122-83-4 | C6H5NOS | 139.172 g/mol

It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is a versatile synthetic intermediate and an active pharmaceutical intermediate.

Solubility
Partly miscible in water.

Notes
Store at room temperature. Keep away from oxidizing agents and heats.
TRUSTED_SUSTAINABILITY

Specifications

CAS 1122-83-4
Color Brown
Density 1.236 g/mL
Boiling Point 200°C
Flash Point 84°C (183°F)
Molecular Formula C6H5NOS
Refractive Index 1.6275
MDL Number MFCD00002084
Quantity 25 g
Beilstein 507682
Synonym n-thionylaniline, n-sulfinylaniline, benzenamine, n-sulfinyl, thionylaniline, phenylthionylamine, phenylsulfinylamine, sulfinylphenylamine, aniline, n-sulfinyl, thionyl imide, phenyl, sulfurous imide, phenyl
Solubility Information Partly miscible in water.
InChI Key FIOJWGRGPONADF-UHFFFAOYSA-N
SMILES C1=CC=C(C=C1)N=S=O
IUPAC Name (sulfinylamino)benzene
Molecular Weight (g/mol) 139.172
PubChem CID 70739
Formula Weight 139.18
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-Thionylaniline
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.