missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ n-Tetradecyltrichlorosilane, 97%

Product Code. 11337465
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11337465 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11337465 Supplier Alfa Aesar™ Supplier No. L05495.03
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Boiling Point 155°C to 157°C (3mmHg)
CAS 18402-22-7
Quantity 1 g
Molecular Formula C14H29Cl3Si
MDL Number MFCD00051440
UN Number UN2987
Beilstein 1764981
Synonym tetradecyltrichlorosilane, trichloro tetradecyl silane, n-tetradecyltrichlorosilane, myristyltrichlorosilane, silane, trichlorotetradecyl, trichloromyristylsilane, trichloro tetradecyl silane #
InChI Key LPMVYGAHBSNGHP-UHFFFAOYSA-N
SMILES CCCCCCCCCCCCCC[Si](Cl)(Cl)Cl
IUPAC Name trichloro(tetradecyl)silane
Molecular Weight (g/mol) 331.821
PubChem CID 87626
Formula Weight 331.83
Percent Purity 97%
Chemical Name or Material n-Tetradecyltrichlorosilane
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.