missing translation for 'onlineSavingsMsg'
Learn More

n-Tetradecane, TRC

Product Code. 30004772
Change view
Click to view available options
Quantity:
2 g
100 g
500 g
Unit Size:
100g
2g
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30004772 500 g 500g
17984835 100 g 100g
17922505 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 30004772 Supplier Toronto Research Chemicals Supplier No. TRCT292250500G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 629-59-4
Molecular Formula C14 H30
Molecular Weight (g/mol) 198.39
Synonym Tetradecane, Cactus Normal Paraffin N 14, Linpar 14, N 14, NSC 72440, Parafol 14-97, Rubitherm RT 2, TS Paraffin TS 4, n-Tetradecane
IUPAC Name tetradecane
SMILES CCCCCCCCCCCCCC

Specifications

Quantity 500 g
InChI Formula InChI=1S/C14H30/c1-3-5-7-9-11-13-14-12-10-8-6-4-2/h3-14H2,1-2H3
Formula Weight 198.2348
Percent Purity >95%
Chemical Name or Material n-Tetradecane
Grade GC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.