missing translation for 'onlineSavingsMsg'
Learn More
Learn More
n-Tetradecane, 99%, Thermo Scientific Chemicals
Click to view available options
Quantity:
100 g
500 g
2500 g
Description
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
n-Tetradecane is used to study the thermodiffusion behavior of the microemulsion droplets of water, n-alkanes and pentaethylene glycol monododecyl ether. It is used as reference material in gas chromatographic analysis, as a solvent and finds applications in organic synthesis.
Solubility
Soluble in ether, acetone and alcohol. Insoluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
Melting Point | 6°C |
Density | 0.763 |
Boiling Point | 252°C to 254°C |
Flash Point | 99°C (211°F) |
Assay Percent Range | 99% |
Odor | Mild |
Linear Formula | CH3(CH2)12CH3 |
Refractive Index | 1.429 |
Quantity | 2500 g |
Solubility Information | Soluble in ether,acetone and alcohol. Insoluble in water. |
Show More |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'