missing translation for 'onlineSavingsMsg'
Learn More

n-Tetracontane, 97% min

Product Code. 11306358
Change view
Click to view available options
Quantity:
0.5 g
2 g
Unit Size:
0.5g
2g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11306358 0.5 g 0.5g
11316358 2 g 2g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.
Product Code. 11306358 Supplier Thermo Scientific Alfa Aesar Supplier No. 011274.02
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy
Regulatory: This product is under excise duties on energy product according to EU directive 2003/96/EEC.

CAS: 4181-95-7 | C40H82 | 563.10 g/mol

n-Tetracontane is used in a passivation layer in copper-phthalocyanine field-effect transistors for improving the charge carrier mobility and used to modify the quasi two-dimensional surface electron system on Au(111).

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 4181-95-7
Molecular Formula C40H82
Molecular Weight (g/mol) 563.10
MDL Number MFCD00015267
InChI Key KUPLEGDPSCCPJI-UHFFFAOYSA-N
Synonym n-tetracontane, unii-i79s9ixb9y, alkane c40, i79s9ixb9y, tetracontane, analytical standard, acmc-209jlw, n-tetracontane 500mg, tetracontane gc
PubChem CID 20149
IUPAC Name tetracontane
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCCC

Specifications

Melting Point 80°C to 84°C
Color White
Density 0.779 g/mL
Boiling Point 150°C (0.001 mmHg)
Linear Formula CH3(CH2)38CH3
Quantity 0.5 g
Beilstein 1800152
Solubility Information Soluble in chloroform. Slightly soluble in methanol.Insoluble in water.
Formula Weight 563.09
Percent Purity ≤97%
Physical Form Flakes
Chemical Name or Material n-Tetracontane
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.