missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylisopropylamine, 98%, Thermo Scientific Chemicals

Catalog Number 11433447
Click to view available options
Quantity:
5 g
25 g
7515-80-2
C7H18N
116.23
MFCD00037059
ZWXQPERWRDHCMZ-UHFFFAOYSA-O
n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine
82023
2-methyl-N-propan-2-ylpropan-2-amine
CC(C)[NH2+]C(C)(C)C
7515-80-2
C7H18N
116.23
MFCD00037059
ZWXQPERWRDHCMZ-UHFFFAOYSA-O
n-tert-butylisopropylamine, 2-propanamine, 2-methyl-n-1-methylethyl, tert-butylisopropylamine, n-isopropyl-2-methylpropan-2-amine, ethanamine, 1,1-dimethyl, n-1-methylethyl, n-isopropyl-tert-butylamine, tert-butyl methylethyl amine, tert-butyl isopropyl amine
82023
2-methyl-N-propan-2-ylpropan-2-amine
CC(C)[NH2+]C(C)(C)C

N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butylisopropylamine, is used as a participates in the synthesis of hindered enamines.

Solubility
It has optimal solubility in water.

Notes
Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions. Keep away from oxidizing agents.
TRUSTED_SUSTAINABILITY
Density 0.727
Boiling Point 98°C
Flash Point −3°C (26°F)
Refractive Index 1.398
Quantity 5 g
UN Number UN2733
Beilstein 1846752
Solubility Information It has optimal solubility in water.
Formula Weight 115.22
Percent Purity 98%
Chemical Name or Material N-tert-Butylisopropylamine
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'