missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butyldimethylsilyl)-N-methyltrifluoroacetamide (Technical Grade), TRC

Product Code. 30336771
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
Product Code. Quantity unitSize
30336771 1 g 1g
1 options
This item is not returnable. View return policy

Product Code. 30336771

Brand: Toronto Research Chemicals TRCB6913101G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material MTBSTFA (with 1% t-BDMCS)
CAS 77377-52-7
Molecular Formula C9 H18 F3 N O Si
Quantity 1 g
Synonym MTBSTFA, N-tert-Butyldimethylsilyl-N-methyltrifluoroacetamide
InChI Formula InChI=1S/C9H18F3NOSi/c1-8(2,3)15(5,6)13(4)7(14)9(10,11)12/h1-6H3
SMILES CN(C(=O)C(F)(F)F)[Si](C)(C)C(C)(C)C
IUPAC Name N-[tert-butyl(dimethyl)silyl]-2,2,2-trifluoro-N-methylacetamide
Molecular Weight (g/mol) 241.33
Formula Weight 241.111
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.