missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butylcyclohexylamine, 97%

Product Code. 11985201 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass bottle
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
11985201 5 g Glass bottle 5g
11975201 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11985201 Supplier Thermo Scientific Acros Supplier No. 439830050

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 51609-06-4 | C10H21N | 155.29 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 51609-06-4
Molecular Formula C10H21N
Molecular Weight (g/mol) 155.29
InChI Key MTEWAFVECQBILW-UHFFFAOYSA-N
Synonym n-tert-butylcyclohexylamine, n-tert-butyl cyclohexanamine, n-t-butylcyclohexylamine, tert-butyl-cyclohexylamine, tert-butylcyclohexyl amine, tert-butyl cyclohexylamine, tert.-butyl-cyclohexyl-amine, n-tertbutyl-n-cyclohexyl amine, n-tert-butyl-n-cyclohexyl amine, n-1,1-dimethylethyl cyclohexanamine
PubChem CID 103920
IUPAC Name N-tert-butylcyclohexanamine
SMILES CC(C)(C)NC1CCCCC1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Color Colorless to Yellow
Density 0.8340g/mL
Boiling Point 172°C to 174°C
Flash Point 50°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Quantity 5 g
Specific Gravity 0.834
Formula Weight 155.29
Percent Purity 97%
Physical Form Liquid
Chemical Name or Material N-tert-Butylcyclohexylamine
Show More Show Less
compliance-icons compliance-icons
Product Identifier
  • N-tert-Butylcyclohexylamine
Signal Word
  • Danger
Hazard Category
  • Flammable liquid Category 3
  • Skin corrosion/irritation Category 1 B
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H226-Flammable liquid and vapour.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.