missing translation for 'onlineSavingsMsg'
Learn More

Alfa Aesar™ N-tert-Butylbenzamide, 98+%

Product Code. p-7044860
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11338205

Brand: Alfa Aesar™ L08509.06

Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Chemical Identifiers

CAS 5894-65-5
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00041337
InChI Key HYWWTHOGCJXTRI-UHFFFAOYSA-N
Synonym n-tert-butyl benzamide, n-t-butylbenzamide, benzamide, n-tert-butyl, benzamide, n-1,1-dimethylethyl, n-t-butyl benzamide, acmc-209m8h, benzamide,1-dimethylethyl
PubChem CID 138622
IUPAC Name N-tert-butylbenzamide
SMILES CC(C)(C)NC(=O)C1=CC=CC=C1

Specifications

Melting Point 133°C to 135°C
Quantity 5 g
Beilstein 2042391
Formula Weight 177.25
Percent Purity ≥98%
Chemical Name or Material N-tert-Butylbenzamide
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.