missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals N-tert-Butylacrylamide, 97%, pure

Product Code. 10463541 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
Packaging:
Glass bottle
Unit Size:
100g
500g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10463541 100 g Glass bottle 100g
10321731 500 g Glass bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10463541 Supplier Thermo Scientific Chemicals Supplier No. 403451000
 Restricted Item
Estimated Shipment: 30-09-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

CAS: 107-58-4 | C7H13NO | 127.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107-58-4
Molecular Formula C7H13NO
Molecular Weight (g/mol) 127.19
MDL Number MFCD00026271
InChI Key XFHJDMUEHUHAJW-UHFFFAOYSA-N
Synonym n-tert-butylacrylamide, tert-butylacrylamide, n-tert-butyl acrylamide, 2-propenamide, n-1,1-dimethylethyl, acrylamide, n-tert-butyl, n-t-butylacrylamide, unii-xj13fsh48k, n-1,1-dimethylethyl-2-propenamide, xj13fsh48k, n-tert-butyl prop-2-enamide
PubChem CID 7877
IUPAC Name N-tert-butylprop-2-enamide
SMILES CC(C)(C)NC(=O)C=C

Specifications

Melting Point 128°C to 130°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula CH2CHCONHC(CH3)3
Quantity 100 g
Formula Weight 127.19
Percent Purity 97%
Grade Pure
Physical Form Chunks and/or Powder
Chemical Name or Material N-tert-Butylacrylamide
Show More Show Less
compliance-icons
Product Identifier
  • N-tert-Butylacrylamide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
Hazard Statement
  • H302-Harmful if swallowed.
Precautionary Statement
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.