missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butyl-alpha-phenylnitrone, 97%

Product Code. p-7023653
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11488027 1 g 1g
11498027 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 11488027 Supplier Thermo Scientific Alfa Aesar Supplier No. A17442.03
Add to basket
This item is not returnable. View return policy

CAS: 3376-24-7 | C11H15NO | 177.247 g/mol

N-tert-Butyl-a-phenylnitrone is a commonly-used free radical trap. It contains radical scavenging activity and an ability to inhibit Cox-2 (cyclooxygenase-2). N-tert-Butyl-a-phenylnitrone is an antioxidant that has been shown to act as a protective agent in several experimental models of neurodegenerative disorders. N-tert-Butyl-a-phenylnitrone inhibits lipid peroxidation in rat liver microsomes. It also prevents the induction of inducible nitric oxide synthase (iNOS) by reactive oxygen species.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
N-tert-Butyl-a-phenylnitrone is a commonly-used free radical trap. It contains radical scavenging activity and an ability to inhibit Cox-2 (cyclooxygenase-2). N-tert-Butyl-a-phenylnitrone is an antioxidant that has been shown to act as a protective agent in several experimental models of neurodegenerative disorders. N-tert-Butyl-a-phenylnitrone inhibits lipid peroxidation in rat liver microsomes. It also prevents the induction of inducible nitric oxide synthase (iNOS) by reactive oxygen species.

Solubility
Soluble in DMSO (10 mg/ml), chloroform (50 mg/ml), and water (20 mg/ml).

Notes
Store in a cool place. Reactive with oxidizing agents.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3376-24-7
Molecular Formula C11H15NO
Molecular Weight (g/mol) 177.247
MDL Number MFCD00008799
InChI Key IYSYLWYGCWTJSG-FMIVXFBMSA-N
Synonym e-n-benzylidene-2-methylpropan-2-amine oxide
PubChem CID 10313352
IUPAC Name N-tert-butyl-1-phenylmethanimine oxide
SMILES CC(C)(C)[N+](=CC1=CC=CC=C1)[O-]

Specifications

Melting Point 71°C to 75°C
Quantity 1 g
Beilstein 2044028
Merck Index 14,7056
Solubility Information Soluble in DMSO (10mg/ml),chloroform (50mg/ml),and water (20mg/ml).
Formula Weight 177.25
Percent Purity 98%
Chemical Name or Material N-tert-Butyl-alpha-phenylnitrone

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.