missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyloxy)succinimide, 98+%

Product Code. 15460778
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15460778 25 g 25g
15440778 1 g 1g
15450778 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15460778 Supplier Thermo Scientific Alfa Aesar Supplier No. H54493.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 13139-12-3 | C9H13NO5 | 215.205 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-12-3
Molecular Formula C9H13NO5
Molecular Weight (g/mol) 215.205
MDL Number MFCD00037903
InChI Key VTGFSVGZCYYHLO-UHFFFAOYSA-N
Synonym boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
PubChem CID 83168
IUPAC Name tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES CC(C)(C)OC(=O)ON1C(=O)CCC1=O

Specifications

Melting Point ∽95°C (decomposition)
Quantity 25 g
Sensitivity Moisture sensitive
Formula Weight 215.2
Percent Purity ≥98%
Chemical Name or Material N-(tert-Butoxycarbonyloxy)succinimide

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.