missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyloxy)succinimide, 97%

Product Code. 10097949 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10097949 25 g 25g
10382110 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10097949 Supplier Thermo Scientific Acros Supplier No. 436240250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

CAS: 13139-12-3 | C9H13NO5 | 215.21 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 13139-12-3
Molecular Formula C9H13NO5
Molecular Weight (g/mol) 215.21
InChI Key VTGFSVGZCYYHLO-UHFFFAOYSA-N
Synonym boc-osu, tert-butyl 2,5-dioxopyrrolidin-1-yl carbonate, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide, tert-butyl succinimido carbonate, 1-tert-butoxycarbonyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-1,1-dimethylethoxy carbonyl oxy, carbonic acid, 1,1-dimethylethyl 2,5-dioxo-1-pyrrolidinyl ester, boc-osu, tert-butyl n-succinimidyl carbonate, n-tert-butoxycarbonyloxy succinimide boc-osu
PubChem CID 83168
IUPAC Name tert-butyl (2,5-dioxopyrrolidin-1-yl) carbonate
SMILES CC(C)(C)OC(=O)ON1C(=O)CCC1=O

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 88°C to 91°C
Color White
Elemental Analysis 48.22 to 52.24% C, 6.25 to 6.77% N
Infrared Spectrum Authentic
Assay Percent Range 97%
Packaging Glass Bottle
Quantity 25 g
Formula Weight 215.21
Percent Purity 97%
Physical Form Powder
Chemical Name or Material N-(tert-Butoxycarbonyloxy)succinimide
Show More Show Less
compliance-icons
Product Identifier
  • N-(tert-Butoxycarbonyloxy)succinimide
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Respiratory sensitiser Category 1
Hazard Statement
  • H317-May cause an allergic skin reaction.
  • H302-Harmful if swallowed.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.