missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-(tert-Butoxycarbonyl)-N-methylethanolamine, TRC
Click to view available options
Quantity:
25 g
Unit Size:
25g
Specifications
Specifications
| Chemical Name or Material | N-(tert-Butoxycarbonyl)-N-methylethanolamine |
| CAS | 57561-39-4 |
| Molecular Formula | C8H17NO3 |
| Quantity | 25 g |
| Synonym | N-Methyl-N-(2-hydroxyethyl)carbamic Acid tert-Butyl Ester, tert-Butyl N-(2-Hydroxyethyl)-N-methylcarbamate, tert-Butyl [2-(Hydroxy)ethyl]methylcarbamate, (2-Hydroxyethyl)(methyl)carbamic Acid tert-Butyl ester, 1,1-Dimethylethyl (2-hydroxyethyl)methylcarbamate, 2-(N-(tert-Butoxycarbonyl)methylamino)ethanol, 2-(N-Boc-N-methylamino)ethanol, 2-(N-tert-Butoxycarbonyl-N-methylamino)ethanol, N-(2-Hydroxyethyl)-N-(methyl)carbamic Acid tert-Butyl Ester, N-(2-Hydroxyethyl)-N-methyl-carbamic Acid tert-Butyl Ester |
| InChI Formula | InChI=1S/C8H17NO3/c1-8(2,3)12-7(11)9(4)5-6-10/h10H,5-6H2,1-4H3 |
| SMILES | OCCN(C(OC(C)(C)C)=O)C |
| IUPAC Name | tert-butyl (2-hydroxyethyl)(methyl)carbamate |
| Molecular Weight (g/mol) | 175.23 |
| Formula Weight | 175.12 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction