missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butoxycarbonyl-L-prolinol, 98%

Product Code. 10135313 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10135313 25 g Glass bottle 25g
10357811 5 g Glass bottle 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10135313 Supplier Thermo Scientific Acros Supplier No. 344560250
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 69610-40-8 | C10H19NO3 | 201.26 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 69610-40-8
Molecular Formula C10H19NO3
Molecular Weight (g/mol) 201.26
MDL Number MFCD00066232
InChI Key BFFLLBPMZCIGRM-QMMMGPOBSA-N
Synonym n-boc-l-prolinol, boc-l-prolinol, s-1-boc-2-pyrrolidinemethanol, s-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, tert-butyl 2s-2-hydroxymethyl pyrrolidine-1-carboxylate, 1-boc-l-prolinol, n-tert-butoxycarbonyl-l-prolinol, boc-prolinol, s---1-boc-2-pyrrolidinemethanol, boc-pro-ol
PubChem CID 643448
IUPAC Name tert-butyl (2S)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
SMILES CC(C)(C)OC(=O)N1CCCC1CO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Melting Point 60°C to 64°C
Color White
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Quantity 25 g
Specific Rotation Condition −52.00 (20.00°C c=1,CHCl3)
Specific Rotation -52
Solubility Information Solubility in water: insoluble in water
Formula Weight 201.26
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butoxycarbonyl-L-prolinol
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.