missing translation for 'onlineSavingsMsg'
Learn More

N-(tert-Butoxycarbonyl)ethanolamine, 98%

Product Code. 10222892 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
Packaging:
Glass Bottle
Unit Size:
25mL
5mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
10222892 25 mL Glass Bottle 25mL
10078801 5 mL Glass Bottle 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 10222892 Supplier Thermo Scientific Acros Supplier No. 303170250
 Restricted Item
In Stock
Add to basket
This item is not returnable. View return policy

CAS: 26690-80-2 | C7H15NO3 | 161.2 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26690-80-2
Molecular Formula C7H15NO3
Molecular Weight (g/mol) 161.2
MDL Number MFCD00056657
InChI Key GPTXCAZYUMDUMN-UHFFFAOYSA-N
Synonym n-boc-ethanolamine, tert-butyl n-2-hydroxyethyl carbamate, boc-glycinol, tert-butyl 2-hydroxyethyl carbamate, n-tert-butoxycarbonyl ethanolamine, boc-ethanolamine, tert-butyl 2-hydroxyethylcarbamate, 2-tert-butoxycarbonylamino-1-ethanol, boc-aminoethanol, n-2-hydroxyethyl carbamic acid tert-butyl ester
PubChem CID 2733206
IUPAC Name tert-butyl N-(2-hydroxyethyl)carbamate
SMILES CC(C)(C)OC(=O)NCCO

Specifications

CAS Min % 97.5
CAS Max % 100.0
Color Colorless to Yellow
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass Bottle
Linear Formula HOCH2CH2NHCO2C(CH3)3
Refractive Index 1.4485 to 1.4505
Density 1.0420g/mL
Quantity 25 mL
Flash Point >110°C
Formula Weight 161.2
Percent Purity 98%
Specific Gravity 1.042
Physical Form Viscous Liquid
Chemical Name or Material N-(tert-Butoxycarbonyl)ethanolamine, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.