missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butoxycarbonyl-D-prolinol, 98%, Thermo Scientific™

Product Code. 10387761 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
1 g
Packaging:
Glass bottle
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging unitSize
10387761 1 g Glass bottle 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10387761 Supplier Thermo Scientific Supplier No. 344550010
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Specifications

CAS 83435-58-9
CAS Min % 97.5
CAS Max % 100.0
Melting Point 59.0°C to 62.0°C
Color White to Beige
Infrared Spectrum Authentic
Assay Percent Range 97.5% min. (GC)
Packaging Glass bottle
Molecular Formula C10H19NO3
MDL Number MFCD00040580
Quantity 1 g
Specific Rotation + 49.00
Synonym boc-d-prolinol, n-boc-d-prolinol, r-tert-butyl 2-hydroxymethyl pyrrolidine-1-carboxylate, r-+-1-boc-2-pyrrolidinemethanol, 1-boc-d-prolinol, boc-d-pro-ol, n-tert-butoxycarbonyl-d-prolinol, n-t-boc-d-prolinol, tert-butyl 2r-2-hydroxymethyl pyrrolidine-1-carboxylate
Solubility Information Solubility in water: insoluble in water.
InChI Key BFFLLBPMZCIGRM-MRVPVSSYSA-N
SMILES CC(C)(C)OC(=O)N1CCCC1CO
IUPAC Name tert-butyl (2R)-2-(hydroxymethyl)pyrrolidine-1-carboxylate
Molecular Weight (g/mol) 201.266
PubChem CID 688279
Formula Weight 201.27
Percent Purity 98%
Physical Form Crystalline Powder
Chemical Name or Material N-tert-Butoxycarbonyl-D-prolinol, 98%
Show More Show Less

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.