missing translation for 'onlineSavingsMsg'
Learn More

N-tert-Butoxycarbonyl-2-piperidinone, TRC

Product Code. 30330322
Change view
Click to view available options
Quantity:
10 g
Unit Size:
10g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30330322 10 g 10g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30330322 Supplier Toronto Research Chemicals Supplier No. TRCB69059510G
 Restricted Item
Estimated Shipment: 02-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material N-tert-Butoxycarbonyl-2-piperidinone
CAS 85908-96-9
Molecular Formula C10H17NO3
Quantity 10 g
Synonym 2-Oxo-1-piperidinecarboxylic Acid 1,1-Dimethylethyl Ester, 1,1-Dimethylethyl 2-Oxopiperidine-1-carboxylate, 2-Oxopiperidine-1-carboxylic Acid tert-Butyl Ester
InChI Formula InChI=1S/C10H17NO3/c1-10(2,3)14-9(13)11-7-5-4-6-8(11)12/h4-7H2,1-3H3
SMILES CC(C)(C)OC(=O)N1CCCCC1=O
IUPAC Name tert-butyl 2-oxopiperidine-1-carboxylate
Molecular Weight (g/mol) 199.25
Formula Weight 199.1208
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.