missing translation for 'onlineSavingsMsg'
Learn More
Learn More
N-t-Boc-2-bromoethylamine (Technical grade, ~90%), TRC
Click to view available options
Quantity:
50 g
Unit Size:
50g
Specifications
Specifications
| Chemical Name or Material | N-t-Boc-2-bromoethylamine |
| CAS | 39684-80-5 |
| Molecular Formula | C7 H14 Br N O2 |
| Quantity | 50 g |
| InChI Formula | InChI=1S/C7H14BrNO2/c1-7(2,3)11-6(10)9-5-4-8/h4-5H2,1-3H3,(H,9,10) |
| SMILES | CC(C)(C)OC(=O)NCCBr |
| IUPAC Name | tert-butyl N-(2-bromoethyl)carbamate |
| Molecular Weight (g/mol) | 224.1 |
| Formula Weight | 223.0208 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction