missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl N-methylcarbamate, 97%

Product Code. 15490319
Change view
Click to view available options
Quantity:
1 g
5 g
25 g
Unit Size:
1g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15490319 25 g 25g
15470319 1 g 1g
15480319 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 15490319 Supplier Thermo Scientific Alfa Aesar Supplier No. H63566.14
Request Bulk or Custom Formats

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 18342-66-0 | C6H8N2O4 | 172.14 g/mol

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18342-66-0
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
MDL Number MFCD00800292
InChI Key XMNGSPOWUCNRMO-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
PubChem CID 4381935
ChEBI CHEBI:83949
IUPAC Name (2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
SMILES CNC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 25 g
Beilstein 1531863
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 172.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Succinimidyl N-methylcarbamate

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.