missing translation for 'onlineSavingsMsg'
Learn More

N-Succinimidyl N-methylcarbamate, 97%, Thermo Scientific Chemicals

Product Code. 15470319
Click to view available options
Quantity:
1 g
5 g
25 g
This item is not returnable. View return policy
This item is not returnable. View return policy

It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
It is an important raw material and intermediate used in organic synthesis, pharmaceuticals, agrochemicals and dyestuff.

Solubility
Slightly soluble in water.

Notes
Moisture Sensitive, store away from moisture. Store in cool. Keep container tightly closed in a dry and well-ventilated place. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 18342-66-0
Molecular Formula C6H8N2O4
Molecular Weight (g/mol) 172.14
MDL Number MFCD00800292
InChI Key XMNGSPOWUCNRMO-UHFFFAOYSA-N
Synonym 2,5-dioxopyrrolidin-1-yl methylcarbamate, n-succinimidyl-n-methylcarbamate, n-succinimidyl n-methylcarbamate, n-succinimdyl-n-methylcarbamate, 2,5-dioxopyrrolidin-1-yl n-methylcarbamate, 1-methylcarbamoyl oxy pyrrolidine-2,5-dione, 2,5-pyrrolidinedione, 1-methylamino carbonyl oxy, mic substitute, methyl isocyanate substitute, methylcarbamic acid succinimidyl ester
PubChem CID 4381935
ChEBI CHEBI:83949
IUPAC Name (2,5-dioxopyrrolidin-1-yl) N-methylcarbamate
SMILES CNC(=O)ON1C(=O)CCC1=O

Specifications

Quantity 1 g
Beilstein 1531863
Sensitivity Moisture sensitive
Solubility Information Slightly soluble in water.
Formula Weight 172.14
Percent Purity 97%
Physical Form Solid
Chemical Name or Material N-Succinimidyl N-methylcarbamate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'